siRNA Details
Gene Symbol: Duox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024141.1 | NP_077055.1 | 5 | 646 | ||
XM_006234862.2 | XP_006234924.1 | 7 | 1610 | |||
GenBank mRNA | AF237962.1 | - | 5 | 646 |
Gene Symbol: Duox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153739.1 | NP_714961.1 | 5 | 656 | ||
XM_006234829.2 | XP_006234891.1 | 6 | 755 | |||
GenBank mRNA | AF542180.1 | - | 5 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
