siRNA Details
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024823.4 | NP_001019994.4 | 7 | 3710 | ||
NM_001159653.1 | NP_001153125.1 | 2379 | ||||
NM_019132.1 | NP_062005.1 | 1639 | ||||
NM_021845.5 | NP_068617.6 | 7 | 3710 | |||
GenBank mRNA | AF107844.1 | - | 2197 | |||
BC061967.1 | - | 1601 | ||||
BC158589.1 | - | 1494 | 9099119 | |||
BC168156.1 | - | 1503 | ||||
FQ214546.1 | - | 1377 | ||||
FQ219804.1 | - | 1069 | ||||
M17525.1 | - | 1639 | ||||
X84047.1 | - | 7 | 2858 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
