siRNA Details
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019387.2 | NP_062260.2 | 3 | 172 | ||
XM_006247908.2 | XP_006247970.1 | 3 | 353 | |||
XM_006247909.2 | XP_006247971.1 | 3 | 353 | |||
GenBank mRNA | AB002811.1 | - | 3 | 178 | ||
AF036344.1 | - | 3 | 208 | |||
BC083561.1 | - | 3 | 172 | 7133877 | ||
U87863.1 | - | 3 | 158 |

More Information
Other Species Targeted

Gene Ontology Categories:
