siRNA Details
Gene Symbol: Defb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022544.2 | NP_071989.1 | 2 | 290 | ||
GenBank mRNA | AF068861.1 | - | 2 | 176 | ||
AY621374.1 | - | 2 | 290 |
Gene Symbol: Defb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037549.1 | NP_001032638.1 | 2 | 152 | ||
GenBank mRNA | AY621340.1 | - | 2 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
