siRNA Details
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023978.2 | NP_076468.2 | 2, 3 | 279 | ||
XM_006239500.2 | XP_006239562.1 | 2, 3 | 731 | |||
XM_006239501.2 | XP_006239563.1 | 2, 3 | 731 | |||
XM_006239502.2 | XP_006239564.1 | 2, 3 | 731 | |||
XM_006239503.2 | XP_006239565.1 | 2, 3 | 730 | |||
XM_006239504.2 | XP_006239566.1 | 2, 3 | 731 | |||
XM_006239505.2 | XP_006239567.1 | 2, 3 | 730 | |||
GenBank mRNA | AB092512.1 | - | 2, 3 | 279 | ||
AB092513.1 | - | 2, 3 | 279 |
