siRNA Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053601.2 | NP_446053.1 | 2, 3 | 275 | ||
GenBank mRNA | CB711943.1 | - | 2, 3 | 211 | ||
FQ213121.1 | - | 2, 3 | 233 | |||
U08290.1 | - | 2, 3 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
