siRNA Details
Gene Symbol: Pitx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053624.2 | NP_446076.1 | 2 | 543 | ||
XM_006253569.2 | XP_006253631.1 | 3 | 361 | |||
GenBank mRNA | AB047298.1 | - | 2 | 564 | ||
BC061966.1 | - | 2 | 543 | 5621094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
