siRNA Details
Gene Symbol: SULT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003167.3 | NP_003158.2 | 2, 3 | 493 | ||
GenBank mRNA | AK289380.1 | - | 2, 3 | 435 | ||
AK313415.1 | - | 2, 3 | 420 | |||
BC020755.1 | - | 2, 3 | 410 | 4733628 | ||
BG563083.1 | - | 2, 3 | 473 | |||
BX509490.1 | - | 2, 3 | 494 | |||
L02337.1 | - | 2, 3 | 418 | |||
L20000.1 | - | 2, 3 | 405 | |||
S43859.1 | - | 2, 3 | 418 | |||
S43861.1 | - | 1, 2 | 187 | |||
U08024.1 | - | 2, 3 | 373 | |||
U08025.1 | - | 2, 3 | 417 | |||
X70222.1 | - | 2, 3 | 405 | |||
X84816.1 | - | 2, 3 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
