siRNA Details
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301285.1 | NP_001288214.1 | 8, 9 | 866 | ||
NM_001301286.1 | NP_001288215.1 | 7, 8 | 690 | |||
XM_006232569.2 | XP_006232631.1 | 8, 9 | 886 | |||
XM_006232570.2 | XP_006232632.1 | 8, 9 | 886 | |||
XM_006232571.2 | XP_006232633.1 | 8, 9 | 886 | |||
XM_006232573.2 | XP_006232635.1 | 8, 9 | 886 | |||
XM_006232575.2 | XP_006232637.1 | 8, 9 | 886 | |||
XM_006232580.2 | XP_006232642.1 | 7, 8 | 790 | |||
XM_006232583.2 | XP_006232645.1 | 7, 8 | 794 | |||
XM_008761124.1 | XP_008759346.1 | 7, 8 | 790 | |||
GenBank mRNA | FQ214881.1 | - | 8, 9 | 864 | ||
FQ215580.1 | - | 8, 9 | 866 | |||
FQ215752.1 | - | 8, 9 | 866 | |||
L24776.1 | - | 690 |

More Information
Other Species Targeted

Gene Ontology Categories:
