siRNA Details
Gene Symbol: Ifi27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203410.1 | NP_981955.1 | 3 | 241 | ||
GenBank mRNA | AF154572.1 | - | 2 | 165 | ||
BC060548.1 | - | 3 | 241 | 6919535 | ||
FQ219860.1 | - | 3 | 299 | |||
FQ220098.1 | - | 3 | 310 | |||
FQ226050.1 | - | 2 | 360 | |||
FQ226998.1 | - | 3 | 274 | |||
FQ230691.1 | - | 2 | 261 | |||
FQ233730.1 | - | 3 | 275 | |||
FQ233981.1 | - | 3 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
