siRNA Details
Gene Symbol: Vegp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053574.1 | NP_446026.1 | 5 | 490 | ||
GenBank mRNA | X74806.1 | - | 5 | 490 |
Gene Symbol: LOC100911507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233718.2 | XP_006233780.1 | 203 |
Gene Symbol: Lcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022945.1 | NP_075234.1 | 5 | 492 | ||
GenBank mRNA | X52016.1 | - | 5 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
