siRNA Details
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053601.2 | NP_446053.1 | 2, 3 | 278 | ||
GenBank mRNA | CB711943.1 | - | 2, 3 | 214 | ||
FQ213121.1 | - | 2, 3 | 236 | |||
U08290.1 | - | 2, 3 | 197 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010923, AK003004, AK160626, BC036984, X83570.
Gene Ontology Categories:
