siRNA Details
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053755.2 | NP_446207.1 | 2 | 262 | ||
GenBank mRNA | AF337809.1 | - | 2 | 267 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053755.2 | NP_446207.1 | 2 | 262 | ||
GenBank mRNA | AF337809.1 | - | 2 | 267 |