siRNA Details
Gene Symbol: Rgcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054008.1 | NP_446460.1 | 2, 3 | 336 | ||
GenBank mRNA | AF036548.1 | - | 2, 3 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
