siRNA Details
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138509.3 | NP_612518.2 | 3 | 226 | ||
XM_006235294.2 | XP_006235356.1 | 3 | 209 | |||
XM_006235295.2 | XP_006235357.1 | 3 | 279 | |||
GenBank mRNA | BC081726.1 | - | 3 | 226 | 7192469 | |
U75920.1 | - | 3 | 194 |
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101000.1 | NP_001094470.1 | 303 | |||
GenBank mRNA | BC105879.1 | - | 303 | 7123955 |
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007656.1 | NP_001007657.1 | 4 | 252 | ||
XM_006239796.2 | XP_006239858.1 | 3 | 180 | |||
XM_008764512.1 | XP_008762734.1 | 3 | 302 | |||
XM_008764513.1 | XP_008762735.1 | 3 | 302 | |||
GenBank mRNA | BC083589.1 | - | 4 | 252 | 7132546 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153058, NM_133350, NM_001162942, NM_001162941, AK028823, and others.
Gene Ontology Categories:
