siRNA Details
Gene Symbol: TCF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003206.3 | NP_003197.2 | 2 | 996 | ||
GenBank mRNA | AF035718.1 | - | 2 | 980 | ||
AF047419.1 | - | 2 | 979 | |||
W73859.1 | - | 2 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
