siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031117.2 | NP_112379.1 | 4 | 496 | ||
GenBank mRNA | FM122235.1 | - | 3 | 292 | ||
M29293.1 | - | 4 | 496 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270712.1 | NP_001257641.1 | 5 | 520 | ||
NM_130738.2 | NP_570094.1 | 4 | 385 | |||
GenBank mRNA | AF101041.1 | - | 4 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
