siRNA Details
Gene Symbol: Ifi27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203410.1 | NP_981955.1 | 4 | 339 | ||
GenBank mRNA | AF154572.1 | - | 3 | 263 | ||
BC060548.1 | - | 4 | 339 | 6919535 | ||
FQ219860.1 | - | 4 | 397 | |||
FQ220098.1 | - | 4 | 408 | |||
FQ226050.1 | - | 3 | 458 | |||
FQ226998.1 | - | 4 | 372 | |||
FQ230691.1 | - | 3 | 359 | |||
FQ233730.1 | - | 4 | 373 | |||
FQ233981.1 | - | 4 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
