siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047893.3 | NP_001041358.2 | 10 | 6512 | ||
XM_006256816.2 | XP_006256878.1 | 10 | 6594 | |||
XM_006256817.2 | XP_006256879.1 | 10 | 6591 | |||
XM_008773126.1 | XP_008771348.1 | 9 | 6477 | |||
XM_008773127.1 | XP_008771349.1 | 6 | 4069 | |||
XM_008773128.1 | XP_008771350.1 | 6 | 5737 | |||
XM_008773129.1 | XP_008771351.1 | 6 | 5719 | |||
XM_008773130.1 | XP_008771352.1 | 6 | 5722 | |||
XM_008773131.1 | XP_008771353.1 | 6 | 4051 | |||
XM_008773132.1 | XP_008771354.1 | 6 | 4054 | |||
GenBank mRNA | AF348365.1 | - | 1 | 484 | ||
BC085701.1 | - | 1 | 820 | |||
FQ211812.1 | - | 6 | 3619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
