siRNA Details
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139194.2 | NP_631933.2 | 3 | 332 | ||
GenBank mRNA | AF104034.1 | - | 3 | 264 | ||
CB810875.1 | - | 3 | 280 | |||
D26113.1 | - | 3 | 357 | |||
FQ233018.1 | - | 3 | 320 |
