siRNA Details
Gene Symbol: THRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190918.1 | NP_001177847.1 | 10 | 2236 | ||
NM_001190919.1 | NP_001177848.1 | 10 | 2334 | |||
NM_003250.5 | NP_003241.2 | 10 | 2353 | |||
GenBank mRNA | BC000261.1 | - | 10 | 2202 | 3501886 | |
BC002728.2 | - | 10 | 2202 | 3628335 | ||
BC035137.1 | - | 10 | 2245 | 5264048 | ||
M24899.1 | - | 10 | 2189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
