siRNA Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173122.3 | NP_775145.2 | 12 | 3421 | ||
XM_006241866.2 | XP_006241928.1 | 11 | 3376 | |||
XM_006241867.2 | XP_006241929.1 | 11 | 3366 | |||
GenBank mRNA | CB578147.1 | - | 4 | 518 | ||
X53232.1 | - | 2 | 164 |

More Information
Other Species Targeted

Gene Ontology Categories:
