siRNA Details
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D13912.1 | - | 13 | 1782 | ||
D29967.1 | - | 1784 | ||||
FQ211106.1 | - | 1797 | ||||
FQ218135.1 | - | 2517 | ||||
M10161.1 | - | 1795 | ||||
M28452.1 | - | 655 | ||||
X96721.1 | - | 1784 |
Gene Symbol: LOC100910877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751127.3 | XP_003751175.1 | 13 | 1812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
