siRNA Details
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003395.2 | NP_001003395.1 | 5, 6 | 758 | ||
NM_001300994.2 | NP_001287923.1 | 4, 5 | 617 | |||
NM_001318903.1 | NP_001305832.1 | 6, 7 | 671 | |||
NM_003287.3 | NP_003278.1 | 5, 6 | 656 | |||
XM_005267122.2 | XP_005267179.1 | 5, 6 | 437 | |||
XM_006715557.3 | XP_006715620.1 | 5, 6 | 902 | |||
XM_017011237.1 | XP_016866726.1 | 5, 6 | 902 | |||
XM_017011238.1 | XP_016866727.1 | 4, 5 | 863 | |||
GenBank mRNA | AK291887.1 | - | 5, 6 | 451 | ||
BM048527.1 | - | 5, 6 | 676 | |||
BM546629.1 | - | 6, 7 | 544 | |||
BQ641312.1 | - | 4, 5 | 496 | |||
U44427.1 | - | 5, 6 | 613 | |||
U44428.1 | - | 2, 3 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
