siRNA Details
Gene Symbol: Vom1r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173298.2 | NP_775420.2 | 1 | 172 | ||
XM_008763052.1 | XP_008761274.1 | 3 | 636 | |||
GenBank mRNA | AY510305.1 | - | 1 | 172 | ||
U36899.1 | - | 3 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
