siRNA Details
Gene Symbol: NR2C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291694.1 | NP_001278623.1 | 5 | 798 | ||
NM_003298.4 | NP_003289.2 | 6 | 855 | |||
XM_011534058.2 | XP_011532360.1 | 9 | 890 | |||
XM_011534059.2 | XP_011532361.1 | 9 | 1796 | |||
XM_011534061.2 | XP_011532363.1 | 7 | 744 | |||
XM_011534063.2 | XP_011532365.1 | 8 | 831 | |||
XM_011534064.2 | XP_011532366.1 | 8 | 993 | |||
XM_011534065.2 | XP_011532367.1 | 7 | 703 | |||
XM_011534066.2 | XP_011532368.1 | 6 | 1506 | |||
XM_017007118.1 | XP_016862607.1 | 7 | 972 | |||
XM_017007119.1 | XP_016862608.1 | 7 | 632 | |||
XM_017007120.1 | XP_016862609.1 | 5 | 640 | |||
GenBank mRNA | AB307708.1 | - | 4 | 436 | ||
AK290590.1 | - | 5 | 798 | |||
BC051670.1 | - | 6 | 628 | 5272493 | ||
BF307171.1 | - | 5 | 454 | |||
HQ995528.1 | - | 5 | 493 | |||
L27586.1 | - | 6 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
