siRNA Details
Gene Symbol: Kcna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173095.3 | NP_775118.1 | 1 | 272 | ||
GenBank mRNA | M26161.1 | - | 1 | 272 | ||
X12589.1 | - | 485 |
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023954.1 | NP_076444.1 | 1 | 1896 | ||
GenBank mRNA | X17621.1 | - | 1 | 680 |
