siRNA Details
Gene Symbol: Brinp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173115.1 | NP_775138.1 | 2 | 1106 | ||
GenBank mRNA | AB077853.1 | - | 2 | 1106 |

More Information
Other Species Targeted

Gene Ontology Categories:
