siRNA Details
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003954.4 | NP_003945.2 | 5 | 736 | ||
XM_011525441.2 | XP_011523743.1 | 6 | 873 | |||
GenBank mRNA | AK290203.1 | - | 5 | 712 | ||
AK313085.1 | - | 5 | 712 | |||
BC035576.1 | - | 5 | 681 | 5497185 | ||
Y10256.1 | - | 4 | 843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
