siRNA Details
Gene Symbol: Vom1r97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173297.2 | NP_775419.2 | 1 | 361 | ||
GenBank mRNA | AY510281.1 | - | 1 | 361 | ||
U36897.1 | - | 1 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
