siRNA Details
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173122.3 | NP_775145.2 | 12 | 4105 | ||
XM_006241866.2 | XP_006241928.1 | 11 | 4060 | |||
XM_006241867.2 | XP_006241929.1 | 11 | 4050 | |||
GenBank mRNA | X53232.1 | - | 2 | 846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
