siRNA Details
Gene Symbol: Testin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173132.2 | NP_775155.1 | 4 | 421 | ||
XM_008771428.1 | XP_008769650.1 | 5 | 781 | |||
GenBank mRNA | U16858.1 | - | 4 | 425 |

More Information
Other Species Targeted

Gene Ontology Categories:
