siRNA Details
Gene Symbol: Defa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033078.1 | NP_001028250.1 | 1 | 113 | ||
XM_006253369.1 | XP_006253431.1 | 2 | 211 | |||
GenBank mRNA | AY623755.1 | - | 1 | 113 |
Gene Symbol: Defa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173329.2 | NP_775451.2 | 2 | 198 | ||
XM_006253367.1 | XP_006253429.1 | 2 | 242 | |||
GenBank mRNA | U16685.1 | - | 2 | 177 | ||
U16686.1 | - | 2 | 198 |
Gene Symbol: Defa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033075.1 | NP_001028247.1 | 1 | 128 | ||
GenBank mRNA | AY623751.1 | - | 1 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
