siRNA Details
Gene Symbol: ZNF132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003433.3 | NP_003424.3 | 3 | 1329 | ||
GenBank mRNA | AK057460.1 | - | 3 | 1329 | ||
AK313895.1 | - | 1 | 811 | |||
BC109107.1 | - | 3 | 1123 | 40035328 | ||
BC109108.1 | - | 3 | 1123 | 40035329 | ||
U09411.1 | - | 1 | 1075 |
