siRNA Details
Gene Symbol: Cyp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175837.1 | NP_787031.1 | 1 | 147 | ||
XM_006238712.2 | XP_006238774.1 | 1 | 159 | |||
GenBank mRNA | BC089761.1 | - | 1 | 125 | 7324500 | |
X07259.1 | - | 1 | 117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
