siRNA Details
Gene Symbol: ZNF136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003437.3 | NP_003428.1 | 3 | 299 | ||
XM_011528269.2 | XP_011526571.1 | 4 | 794 | |||
XM_017027243.1 | XP_016882732.1 | 2 | 557 | |||
GenBank mRNA | AK123008.1 | - | 2 | 1316 | ||
AK291242.1 | - | 3 | 294 | |||
AK299601.1 | - | 2 | 122 | |||
BC006421.1 | - | 3 | 252 | 4040430 | ||
DB200139.1 | - | 3 | 299 | |||
U09367.1 | - | 3 | 223 |
