siRNA Details
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177931.2 | NP_808792.1 | 1 | 86 | ||
XM_006238515.2 | XP_006238577.1 | 2 | 288 | |||
XM_006238516.2 | XP_006238578.1 | 3 | 401 | |||
XM_006238518.2 | XP_006238580.1 | 2 | 288 | |||
GenBank mRNA | AB105378.1 | - | 1 | 86 |
