siRNA Details
Gene Symbol: Il1rapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177935.2 | NP_808796.1 | 1, 2 | 89 | ||
GenBank mRNA | AY216593.1 | - | 1, 2 | 89 |

More Information
Other Species Targeted

Gene Ontology Categories:
