siRNA Details
Gene Symbol: LOC100359767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751626.2 | XP_003751674.1 | 2 | 216 | ||
XM_006253378.1 | XP_006253440.1 | 2 | 224 |
Gene Symbol: Defa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173329.2 | NP_775451.2 | 2 | 204 | ||
XM_006253367.1 | XP_006253429.1 | 2 | 248 | |||
GenBank mRNA | U16685.1 | - | 2 | 183 | ||
U16686.1 | - | 2 | 204 |
Gene Symbol: Defa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033075.1 | NP_001028247.1 | 1 | 134 | ||
GenBank mRNA | AY623751.1 | - | 1 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
