siRNA Details
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 2 | 161 | ||
XM_006237141.2 | XP_006237203.1 | 3 | 684 | |||
XM_006237142.2 | XP_006237204.1 | 2 | 1740 | |||
XM_006237143.2 | XP_006237205.1 | 2 | 538 | |||
XM_006237145.2 | XP_006237207.1 | 3 | 1001 | |||
XM_008763224.1 | XP_008761446.1 | 3 | 1002 | |||
XM_008763225.1 | XP_008761447.1 | 4 | 755 | |||
XM_008763226.1 | XP_008761448.1 | 2 | 855 | |||
GenBank mRNA | AF315467.1 | - | 2 | 475 | ||
BC078692.1 | - | 2 | 253 | |||
BC162051.1 | - | 2 | 161 | 7112072 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081317, AK012639, AK089032, BC048508.
Gene Ontology Categories:
