siRNA Details
Gene Symbol: Olr1361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173333.1 | NP_775455.1 | 1 | 249 | ||
GenBank mRNA | M64377.1 | - | 1 | 249 |
Gene Symbol: Olr1358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214822.1 | NP_999987.1 | 1 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
