siRNA Details
Gene Symbol: Olr1082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173335.1 | NP_775457.1 | 1 | 167 | ||
GenBank mRNA | M64381.1 | - | 1 | 167 |
Gene Symbol: Olr1079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000711.1 | NP_001000711.1 | 1 | 167 |
Gene Symbol: Olr1081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000592.1 | NP_001000592.1 | 1 | 167 |
Gene Symbol: Olr1083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000710.1 | NP_001000710.1 | 1 | 167 |
Gene Symbol: Olr1085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000420.1 | NP_001000420.1 | 1 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
