siRNA Details
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175756.1 | NP_786932.1 | 3 | 209 | ||
XM_006250233.1 | XP_006250295.1 | 3 | 226 | |||
XM_006250234.2 | XP_006250296.1 | 4 | 239 | |||
XM_006250235.1 | XP_006250297.1 | 3 | 223 | |||
GenBank mRNA | BC091113.1 | - | 3 | 203 | 7323350 | |
EF424788.1 | - | 3 | 178 | |||
FQ228752.1 | - | 3 | 228 | |||
X73371.1 | - | 3 | 243 |
