siRNA Details
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175764.2 | NP_786940.1 | 3 | 376 | ||
XM_006236637.2 | XP_006236699.1 | 5 | 640 | |||
XM_006236639.2 | XP_006236701.1 | 4 | 597 | |||
GenBank mRNA | FQ222223.1 | - | 4 | 612 | ||
X59993.1 | - | 3 | 376 |
