siRNA Details
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 4, 5 | 462 | ||
XM_006237141.2 | XP_006237203.1 | 5, 6 | 985 | |||
XM_006237142.2 | XP_006237204.1 | 4, 5 | 2041 | |||
XM_006237143.2 | XP_006237205.1 | 4, 5 | 839 | |||
XM_006237145.2 | XP_006237207.1 | 5, 6 | 1302 | |||
XM_006237146.1 | XP_006237208.1 | 2, 3 | 143 | |||
XM_008763224.1 | XP_008761446.1 | 5, 6 | 1303 | |||
XM_008763225.1 | XP_008761447.1 | 6, 7 | 1056 | |||
XM_008763226.1 | XP_008761448.1 | 4, 5 | 1156 | |||
XM_008763227.1 | XP_008761449.1 | 2, 3 | 270 | |||
GenBank mRNA | AF315467.1 | - | 4, 5 | 776 | ||
BC078692.1 | - | 4, 5 | 554 | |||
BC162051.1 | - | 4, 5 | 462 | 7112072 |

More Information
Other Species Targeted

Gene Ontology Categories:
