siRNA Details
Gene Symbol: Cacna2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175592.2 | NP_783182.1 | 25 | 2126 | ||
XM_006243734.2 | XP_006243796.1 | 25 | 2400 | |||
XM_006243735.2 | XP_006243797.1 | 25 | 2400 | |||
XM_006243736.2 | XP_006243798.1 | 24 | 2379 | |||
XM_006243737.2 | XP_006243799.1 | 24 | 2379 | |||
GenBank mRNA | AF486277.1 | - | 25 | 2126 |
