siRNA Details
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233454.2 | XP_006233516.1 | 14 | 2948 | ||
XM_006233455.2 | XP_006233517.1 | 14 | 3097 | |||
XM_006233458.2 | XP_006233520.1 | 11 | 3078 | |||
XM_008761498.1 | XP_008759720.1 | 13 | 3003 | |||
GenBank mRNA | AF532970.1 | - | 13 | 2848 |
