siRNA Details
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177928.3 | NP_808789.1 | 2 | 164 | ||
GenBank mRNA | AB081730.1 | - | 2 | 139 | ||
AY831728.1 | - | 2 | 137 | |||
BC085681.1 | - | 2 | 164 | 7192109 |

More Information
Other Species Targeted

Gene Ontology Categories:
