siRNA Details
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 3 | 373 | ||
NM_001271536.1 | NP_001258465.1 | 3 | 459 | |||
NM_012495.2 | NP_036627.1 | 3 | 475 | |||
XM_006230209.2 | XP_006230271.1 | 5 | 721 | |||
XM_006230210.2 | XP_006230272.1 | 4 | 631 | |||
XM_006230211.1 | XP_006230273.1 | 4 | 486 | |||
GenBank mRNA | BC064440.1 | - | 3 | 452 | 5623628 | |
CF975930.1 | - | 3 | 433 | |||
CO398643.1 | - | 3 | 366 | |||
FQ212782.1 | - | 3 | 426 | |||
FQ213869.1 | - | 3 | 456 | |||
FQ214510.1 | - | 3 | 428 | |||
FQ216189.1 | - | 3 | 381 | |||
FQ216820.1 | - | 3 | 377 | |||
FQ224221.1 | - | 3 | 466 | |||
FQ228954.1 | - | 3 | 405 | |||
FQ229100.1 | - | 3 | 420 | |||
M12919.1 | - | 3 | 475 | |||
M14420.1 | - | 3 | 373 |
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013943.1 | NP_001013965.1 | 1 | 372 | ||
GenBank mRNA | BC079243.1 | - | 1 | 372 | 7132739 |

More Information
Other Species Targeted

Gene Ontology Categories:
