siRNA Details
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012514.1 | NP_036646.1 | 4 | 294 | ||
XM_008768092.1 | XP_008766314.1 | 4 | 510 | |||
GenBank mRNA | AF036760.1 | - | 4 | 294 | ||
U60523.1 | - | 3 | 158 |
